3-Fluorbenzaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11396.09
A11396.18
A11396.30
A11396.09EA
30.1
EUR
A11396.09
A11396.18
A11396.30
3-Fluorbenzaldehyd ≥97%
3-Fluorbenzaldehyd
Purity: 97%
CAS No.: 456-48-4
Molecular Formula: C7H5FO
Formula Weight: 124.11
Flash Point: 56°(132°F)
Refractive Index: 1.5190
Boiling Point: 173°
Beilstein Reference: 970178
UN No.: 1989
MDL No.: MFCD00003348
Formel:
C₇H₅FO Siedepunkt: 173 °C Dichte: 1,174 Flammpunkt: 56 °C (132 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003348 CAS-Nummer: 456-48-4 EINECS: 207-266-9 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...