Neocuproin Hemihydrat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,9-Dimethyl-1,10-phenanthrolin Hemihydrat

A11398.06 A11398.14 A11398.22
A11398.06EA 58.7 EUR
A11398.06 A11398.14 A11398.22
Neocuproin Hemihydrat ≥98%
Neocuproin Hemihydrat

Purity: 98+% CAS No.: 34302-69-7 Molecular Formula: C14H12N2.0.5H2O Formula Weight: 217.27 (208.26anhy) Melting Point: 160-164° Beilstein Reference: 153179 Merck Reference: 14,6449 MDL No.: MFCD00149306

Formel: C₂₈H₂₆N₄O
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149306
CAS-Nummer: 34302-69-7
EINECS: 207-601-9
Merck Index: 14,6449

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR