α,α,α-Trifluoracetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98%
CAS No.: 434-45-7
Molecular Formula: C8H5F3O
Formula Weight: 174.12
Flash Point: 42°(108°F)
Refractive Index: 1.4595
Melting Point: -40°
Boiling Point: 165-166°
Beilstein Reference: 1866286
UN No.: 1224
MDL No.: MFCD00000420
Formel:
C₈H₅F₃O Siedepunkt: 165…166 °C Schmelzpunkt: –40 °C Dichte: 1,240 Flammpunkt: 42 °C (108 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000420 CAS-Nummer: 434-45-7 EINECS: 207-103-1 UN: 1224 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...