(4-Bromphenyl)methylsulfon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Brom-4-(methylsulfonyl)benzol
A11427.09
A11427.18
A11427.09EA
47
EUR
A11427.09
A11427.18
(4-Bromphenyl)methylsulfon ≥98%
(4-Bromphenyl)methylsulfon
Purity: 98+%
CAS No.: 3466-32-8
Molecular Formula: C7H7BrO2S
Formula Weight: 235.10
Melting Point: 101-102°
Beilstein Reference: 2045675
Formel:
C₇H₇BrO₂S Molecular Weight: 235.1 g/mol Schmelzpunkt: 101…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025065 CAS-Nummer: 3466-32-8 EINECS: 222-421-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...