1,3-Phenylendiessigsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A11429.14 A11429.04 A11429.09
A11429.14EA 252 EUR
A11429.14 A11429.04 A11429.09
1,3-Phenylendiessigsäure ≥97%
1,3-Phenylendiessigsäure

Purity: 97% CAS No.: 19806-17-8 Molecular Formula: C10H10O4 Formula Weight: 194.19 Odour: Odourless Melting Point: 172-178° Beilstein Reference: 2616749 MDL No.: MFCD00004341

Formel: C₁₀H₁₀O₄
Schmelzpunkt: 172…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004341
CAS-Nummer: 19806-17-8
EINECS: 243-332-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR