p-Toluenesulphinic acid sodium salt, (max. 5% H₂O) 97% (Trockengewicht)

Lieferant: Thermo Fisher Scientific
Achtung

A11430.14 A11430.22 A11430.36
A11430.14EA 33.4 EUR
A11430.14 A11430.22 A11430.36
p-Toluenesulphinic acid sodium salt, (max. 5% H₂O) 97% (Trockengewicht)
p-Toluenesulphinic acid sodium salt

Purity: 97% CAS No.: 824-79-3 Molecular Formula: C7H7NaO2S Formula Weight: 178.19 Melting Point: 300° Beilstein Reference: 4621454 MDL No.: MFCD00149640

Formel: C₇H₇NaO₂S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149640
CAS-Nummer: 824-79-3
EINECS: 212-538-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR