2,5-Dihydroxy-p-benzochinon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,5-Dihydroxy-1,4-benzoquinone

A11442.06 A11442.14 A11442.18
A11442.06EA 30.4 EUR
A11442.06 A11442.14 A11442.18
2,5-Dihydroxy-p-benzochinon ≥98%
2,5-Dihydroxy-p-benzochinon

Purity: 98% CAS No.: 615-94-1 Molecular Formula: C6H4O4 Formula Weight: 140.10 Melting Point: ca 220° subl. Beilstein Reference: 1939229

Formel: C₆H₄O₄
Molecular Weight: 140.1 g/mol
Schmelzpunkt: ∼ 220 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001598
CAS-Nummer: 615-94-1
EINECS: 210-454-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR