DL-Methionin ≥99%
Lieferant: Thermo Fisher Scientific
A11457.0B
A11457.0I
A11457.30
A11457.0BEA
84.4
EUR
A11457.0B
A11457.0I
A11457.30
DL-Methionin ≥99%
DL-Methionin
Purity: 99%
CAS No.: 59-51-8
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Melting Point: ca 272° dec.
Beilstein Reference: 636185
Merck Reference: 14,5975
MDL No.: MFCD00063096
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...