3-Amino-2-cyclohexen-1-on, (max. 5% H₂O) 98% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11474.06
A11474.14
A11474.06EA
22.3
EUR
A11474.06
A11474.14
3-Amino-2-cyclohexen-1-on, (max. 5% H₂O) 98% (Trockengewicht)
3-Amino-2-cyclohexen-1-on
Purity: 98%
CAS No.: 5220-49-5
Molecular Formula: C6H9NO
Formula Weight: 111.15
Melting Point: 128-133°(dried)
Beilstein Reference: 471354
Formel:
C₆H₉NO Siedepunkt: 211 °C (1013 hPa) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013783 CAS-Nummer: 5220-49-5 EINECS: 226-014-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...