3-Butin-1-ol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
But-3-in-1-ol, 1-Hydroxy-3-butin, 4-Hydroxy-1-butin
A11477.06
A11477.14
A11477.30
A11477.06EA
40.1
EUR
A11477.06
A11477.14
A11477.30
3-Butin-1-ol ≥98%
3-Butin-1-ol
Purity: 98+%
CAS No.: 927-74-2
Molecular Formula: C4H6O
Formula Weight: 70.09
Flash Point: 36°(96°F)
Refractive Index: 1.4405
Odour: Characteristic
Melting Point: -63°
Boiling Point: 129-130°
Beilstein Reference: 773710
UN No.: 1987
MDL No.: MFCD00002955
Formel:
C₄H₆O Molecular Weight: 70.09 g/mol Siedepunkt: 129…130 °C Schmelzpunkt: –63 °C Dichte: 0.927 Flammpunkt: 36 °C (96 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002955 CAS-Nummer: 927-74-2 EINECS: 213-161-9 UN: 1987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...