3-Iod-p-toluidin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A11485.06 A11485.14 A11485.22
A11485.06EA 35.3 EUR
A11485.06 A11485.14 A11485.22
3-Iod-p-toluidin ≥98%
3-Iod-p-toluidin

Purity: 98% CAS No.: 35944-64-0 Molecular Formula: C7H8IN Formula Weight: 233.05 Melting Point: 34-37° Beilstein Reference: 2353567 UN No.: 2811 MDL No.: MFCD00047843

Formel: C₇H₈IN
Schmelzpunkt: 34…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047843
CAS-Nummer: 35944-64-0
EINECS: 252-803-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR