Thymidin (2'-Deoxythymidin) ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
Purity: 99%
CAS No.: 50-89-5
Molecular Formula: C10H14N2O5
Formula Weight: 242.23
Physical Form: Powder
Appearance: White
Odour: Odourless
Melting Point: 186-190°
Beilstein Reference: 89285
Merck Reference: 14,9397
Optical Rotation: +19° (c=1 in water)
MDL No.: MFCD00006537
Formel:
C₁₀H₁₄N₂O₅ Molecular Weight: 242.23 g/mol Schmelzpunkt: 186…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006537 CAS-Nummer: 50-89-5 EINECS: 200-070-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...