trans-2-Hexenal 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Blätteraldehyd, (E)-2-Hexenal

A11513.14 A11513.22
A11513.14EA 38 EUR
A11513.14 A11513.22
trans-2-Hexenal 96%
trans-2-Hexenal

Purity: 96% CAS No.: 6728-26-3 Molecular Formula: C6H10O Formula Weight: 98.15 Storage Temperature: 2°C to 8°C Flash Point: 38°(100°F) Refractive Index: 1.4470 Odour: Sweet Boiling Point: 37-38°/8mm Beilstein Reference: 1699684 UN No.: 1988 MDL No.: MFCD00007008

Formel: C₆H₁₀O
Molecular Weight: 98.14 g/mol
Siedepunkt: 37…38 °C (8 mmHg)
Dichte: 0.848
Flammpunkt: 38 °C (100 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00007008
CAS-Nummer: 6728-26-3
EINECS: 229-778-1
UN: 1988
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR