Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
A11534.06
A11534.14
A11534.06EA
40.2
EUR
A11534.06
A11534.14
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat 98%
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Purity: 98%
CAS No.: 31037-02-2
Molecular Formula: C7H11N3O2
Formula Weight: 169.18
Melting Point: 97-102°
Beilstein Reference: 131200
Formel:
C₇H₁₁N₃O₂ Schmelzpunkt: 97…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051652 CAS-Nummer: 31037-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...