Methacrylsäure ≥99% stabilisiert
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Methylpropensäure
Stabilisierung:
Stabilisiert mit Hydrochinon oder Hydrochinonmonomethylether
Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly(methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.
Purity: 99%
CAS No.: 79-41-4
Molecular Formula: C4H6O2
Formula Weight: 86.09
Flash Point: 65°(149°F)
Refractive Index: 1.4310
Odour: Acrid
pH: 2.0-2.2
Melting Point: 16°
Boiling Point: 163°
Beilstein Reference: 1719937
Merck Reference: 14,5941
UN No.: 2531
MDL No.: MFCD00002651
Formel:
C₄H₆O₂ Molecular Weight: 86.09 g/mol Siedepunkt: 163 °C Schmelzpunkt: 16 °C Dichte: 1.015 Flammpunkt: 65 °C (149 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002651 CAS-Nummer: 79-41-4 EINECS: 201-204-4 UN: 2531 ADR: 8 II Merck Index: 14,5941 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...