α-Chlor-2-fluortoluol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(Chlormethyl)-2-fluorbenzol, 2-Fluorbenzylchlorid, α-Chlor-o-fluortoluol

A11566.14 A11566.22
A11566.14EA 48.2 EUR
A11566.14 A11566.22
α-Chlor-2-fluortoluol ≥98%
α-Chlor-2-fluortoluol

Purity: 98+% CAS No.: 345-35-7 Molecular Formula: C7H6ClF Formula Weight: 144.58 Flash Point: 57°(134°F) Refractive Index: 1.5154 Odour: Pungent Boiling Point: 175° Beilstein Reference: 471699 UN No.: 2920 MDL No.: MFCD00000892

Formel: C₇H₆ClF
Molecular Weight: 144.58 g/mol
Siedepunkt: 175 °C
Dichte: 1.216
Flammpunkt: 57 °C (134 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000892
CAS-Nummer: 345-35-7
EINECS: 206-460-0
UN: 2920
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR