2-Propylthiazol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11574.06
A11574.14
A11574.06EA
73
EUR
A11574.06
A11574.14
2-Propylthiazol ≥99%
2-Propylthiazole
Purity: 99%
CAS No.: 17626-75-4
Molecular Formula: C6H9NS
Formula Weight: 127.20
Flash Point: 54°(129°F)
Refractive Index: 1.5055
Boiling Point: 62°/15mm
UN No.: 1993
MDL No.: MFCD00053154
Formel:
C₆H₉NS Molecular Weight: 127.20 g/mol Siedepunkt: 62 °C (15 mmHg) Dichte: 1.03 Flammpunkt: 54 °C (129 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053154 CAS-Nummer: 17626-75-4 EINECS: 241-606-7 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...