2-Fluor-6-iodbenzonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11576.06
A11576.06EA
71.6
EUR
A11576.06
2-Fluor-6-iodbenzonitril ≥98%
2-Fluor-6-iodbenzonitril
Purity: 98%
CAS No.: 79544-29-9
Molecular Formula: C7H3FIN
Formula Weight: 247.01
Melting Point: 48-52°
Beilstein Reference: 4971873
UN No.: 3439
MDL No.: MFCD00015478
Formel:
C₇H₃FIN Molecular Weight: 247.01 g/mol Siedepunkt: 281 Schmelzpunkt: 48…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015478 CAS-Nummer: 79544-29-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...