2-Fluorpyridin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11627.09
A11627.14
A11627.22
A11627.30
A11627.09EA
43
EUR
A11627.09
A11627.14
A11627.22
A11627.30
2-Fluorpyridin ≥99%
2-Fluoropyridine
Purity: 99%
CAS No.: 372-48-5
Molecular Formula: C5H4FN
Formula Weight: 97.09
Flash Point: 24°(75°F)
Refractive Index: 1.4670
Boiling Point: 124-126°
Beilstein Reference: 1515
UN No.: 1993
MDL No.: MFCD00006224
Formel:
C₅H₄FN Siedepunkt: 124…126 °C Dichte: 1,129 Flammpunkt: 24 °C (75 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006224 CAS-Nummer: 372-48-5 EINECS: 206-757-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...