3-(3,4-Dihydroxyphenyl)propionsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydrocaffeic acid

A11638.06 A11638.14 A11638.22
A11638.06EA 54.6 EUR
A11638.06 A11638.14 A11638.22
3-(3,4-Dihydroxyphenyl)propionsäure ≥98%
3-(3,4-Dihydroxyphenyl)propionsäure

Purity: 98+% CAS No.: 1078-61-1 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 136-140° Beilstein Reference: 2213449

Formel: C₉H₁₀O₄
Molecular Weight: 182.18 g/mol
Schmelzpunkt: 136…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002776
CAS-Nummer: 1078-61-1
EINECS: 214-083-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR