Leishman's Eosin-Methylenblau Färbemittel für Hämatologie
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Leishmans Eosin-Methylenblau, Leishman's Farbstoff
A11644.14
A11644.14EA
40.4
EUR
A11644.14
Leishman's Eosin-Methylenblau Färbemittel für Hämatologie
Leishman's Eosin-Methylenblau
Leishman stains are designed to differentiate and identify leukocytes.
- Incompatible with oxidising agents
- Soluble in water, methanol, ethanol, and methyl cellosolve
- Acts like fluorescent stains for the detection of malaria parasites
Used to identify parasites, leukocytes, trypanosomas in hematology and tropical medicine applications. Can also be used for bone marrow staining.
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00131498 CAS-Nummer: 12627-53-1 EINECS: 235-732-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...