di-Natriumoxalat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A11648.0E A11648.22 A11648.36
A11648.0EEA 299 EUR
A11648.0E A11648.22 A11648.36
di-Natriumoxalat ≥99%
di-Natriumoxalat

Purity: 99% CAS No.: 62-76-0 Molecular Formula: Na2C2O4 Formula Weight: 134.00 Odour: Odourless Melting Point: ca 250-270° dec. Beilstein Reference: 3631622 Merck Reference: 14,8650 MDL No.: MFCD00012465

Formel: Na₂C₂O₄
Dichte: 2,340
Lagertemperatur: Raumtemperatur
MDL: MFCD00012465
CAS-Nummer: 62-76-0
EINECS: 200-550-3
Merck Index: 14,8650

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR