Caprinaldehyd 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Decanal, Decylaldehyd

A11656.AE A11656.AP
A11656.AEEA 30.4 EUR
A11656.AE A11656.AP
Caprinaldehyd 96%
Caprinaldehyd

Purity: 96% CAS No.: 112-31-2 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 85°(185°F) Refractive Index: 1.4280 Melting Point: -5° Boiling Point: 207-209° Beilstein Reference: 1362530 MDL No.: MFCD00007031

Formel: C₁₀H₂₀O
Molecular Weight: 156.27 g/mol
Siedepunkt: 207…209 °C
Schmelzpunkt: –5 °C
Dichte: 0.828
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007031
CAS-Nummer: 112-31-2
EINECS: 203-957-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR