Fluorescein Dinatriumsalz Hydrat

Lieferant: Thermo Fisher Scientific

A11659.0E A11659.22 A11659.36
A11659.0EEA 332 EUR
A11659.0E A11659.22 A11659.36
Fluorescein Dinatriumsalz Hydrat
Fluorescein Dinatriumsalz Hydrat

Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.


CAS No.: 518-47-8 Molecular Formula: C20H10Na2O5.xH2O Formula Weight: 376.28 Odour: Odourless pH: 8.3 Melting Point: 250° Beilstein Reference: 3833041 Merck Reference: 14,4159 MDL No.: MFCD00167039

Formel: C₂₀H₁₀Na₂O₅·xH₂O
Schmelzpunkt: >250 °C
Dichte: 1,601 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00167039
CAS-Nummer: 518-47-8
EINECS: 208-253-0
Merck Index: 14,4159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR