Fluorescein Dinatriumsalz Hydrat
Lieferant: Thermo Fisher Scientific
Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.
CAS No.: 518-47-8
Molecular Formula: C20H10Na2O5.xH2O
Formula Weight: 376.28
Odour: Odourless
pH: 8.3
Melting Point:
Formel:
C₂₀H₁₀Na₂O₅·xH₂O Schmelzpunkt: >250 °C Dichte: 1,601 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167039 CAS-Nummer: 518-47-8 EINECS: 208-253-0 Merck Index: 14,4159 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...