Bis(trimethylsilyl)carbodiimid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11670.06
A11670.14
A11670.06EA
84.9
EUR
A11670.06
A11670.14
Bis(trimethylsilyl)carbodiimid ≥97%
Bis(trimethylsilyl)carbodiimid
Purity: 97%
CAS No.: 1000-70-0
Molecular Formula: C7H18N2Si2
Formula Weight: 186.41
Flash Point: 42°(107°F)
Refractive Index: 1.4340
Odour: Mild
Melting Point: -23°
Boiling Point: 163-164°
Beilstein Reference: 2042082
UN No.: 1993
MDL No.: MFCD00051538
Formel:
C₇H₁₈N₂Si₂ Molecular Weight: 186.4 g/mol Siedepunkt: 163…164 °C Schmelzpunkt: –23 °C Dichte: 0.823 Flammpunkt: 42 °C (107 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051538 CAS-Nummer: 1000-70-0 EINECS: 213-673-2 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...