Acetylcholiniodid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Acetoxyethyl)trimethylammoniumiodid, Acetylcholiniumiodid

A11694.14
A11694.14EA 75.5 EUR
A11694.14
Acetylcholiniodid ≥98%
Acetylcholiniodid

Purity: 98% CAS No.: 2260-50-6 Molecular Formula: CH3CO2CH2CH2N(CH3)3I Formula Weight: 273.11 Storage Temperature: 2°C to 8°C Melting Point: 161-165° Beilstein Reference: 3571339 MDL No.: MFCD00011815

Formel: H₃CC(O)O(CH₂)₂N(CH₃)₃I
Molecular Weight: 273,11 g/mol
Schmelzpunkt: 161…165 °C
Dichte: 1,67 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00011815
CAS-Nummer: 2260-50-6
EINECS: 218-862-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR