2'-Methylacetophenon ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Methyl-o-tolylketon, o-Methylacetophenon
A11695.06
A11695.14
A11695.22
A11695.06EA
24.5
EUR
A11695.06
A11695.14
A11695.22
2'-Methylacetophenon ≥98%
2'-Methylacetophenon
Purity: 98%
CAS No.: 577-16-2
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 75°(167°F)
Refractive Index: 1.5318
Boiling Point: 214°
Beilstein Reference: 907005
MDL No.: MFCD00008734
Formel:
C₉H₁₀O Molecular Weight: 134.18 g/mol Siedepunkt: 214 °C Dichte: 1.020 Flammpunkt: 75 °C (167 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008734 CAS-Nummer: 577-16-2 EINECS: 209-408-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...