Methyl-4-aminobenzoat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11715.14
A11715.22
A11715.36
A11715.14EA
59.7
EUR
A11715.14
A11715.22
A11715.36
Methyl-4-aminobenzoat 98%
Methyl-4-aminobenzoat
Purity: 98%
CAS No.: 619-45-4
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 110-114°
Beilstein Reference: 775913
Formel:
C₈H₉NO₂ Siedepunkt: 299 °C (1013 hPa) Schmelzpunkt: 110…114 °C Dichte: 1,216 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007891 CAS-Nummer: 619-45-4 EINECS: 210-598-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...