Diethylamin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A11716.0F A11716.AE A11716.AP
A11716.0FEA 68 EUR
A11716.0F A11716.AE A11716.AP
Diethylamin ≥99%
Diethylamin

Purity: 99+% CAS No.: 109-89-7 Molecular Formula: C4H11N Formula Weight: 73.14 Flash Point: -28°(-18°F) Refractive Index: 1.3850 Odour: Ammonia-like Melting Point: -50° Boiling Point: 54-55° Beilstein Reference: 605268 Merck Reference: 14,3111 UN No.: 1154 MDL No.: MFCD00009032

Formel: (C₂H₅)₂NH
Siedepunkt: 54…55 °C
Schmelzpunkt: –50 °C
Dichte: 0,706
Flammpunkt: –28 °C (–18 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009032
CAS-Nummer: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 14,3111
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR