Diethylamin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 109-89-7
Molecular Formula: C4H11N
Formula Weight: 73.14
Flash Point: -28°(-18°F)
Refractive Index: 1.3850
Odour: Ammonia-like
Melting Point: -50°
Boiling Point: 54-55°
Beilstein Reference: 605268
Merck Reference: 14,3111
UN No.: 1154
MDL No.: MFCD00009032
Formel:
(C₂H₅)₂NH Siedepunkt: 54…55 °C Schmelzpunkt: –50 °C Dichte: 0,706 Flammpunkt: –28 °C (–18 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009032 CAS-Nummer: 109-89-7 EINECS: 203-716-3 UN: 1154 ADR: 3,II Merck Index: 14,3111 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...