3'-Fluoracetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
m-Fluoracetophenon
A11733.06
A11733.14
A11733.22
A11733.06EA
57
EUR
A11733.06
A11733.14
A11733.22
3'-Fluoracetophenon ≥98%
3'-Fluoracetophenon
Purity: 98+%
CAS No.: 455-36-7
Molecular Formula: C8H7FO
Formula Weight: 138.14
Flash Point: 80°(176°F)
Refractive Index: 1.5090
Melting Point: -3°
Boiling Point: 80-81°/9mm
Beilstein Reference: 907041
MDL No.: MFCD00000338
Formel:
C₈H₇FO Molecular Weight: 138.14 g/mol Siedepunkt: 80…81 °C (9 mmHg) Schmelzpunkt: –3 °C Dichte: 1.130 Flammpunkt: 80 °C (176 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000338 CAS-Nummer: 455-36-7 EINECS: 207-245-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...