trans-2-Hydroxyzimtsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A11740.14 A11740.22
A11740.14EA 76.5 EUR
A11740.14 A11740.22
trans-2-Hydroxyzimtsäure ≥98%
trans-2-Hydroxyzimtsäure

Purity: 98+% CAS No.: 614-60-8 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: ca 210° dec. Beilstein Reference: 1100900

Formel: C₉H₈O₃
Siedepunkt: 330 °C (1013 hPa)
Schmelzpunkt: ∼ 210 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004379
CAS-Nummer: 614-60-8
EINECS: 210-386-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR