(4-Biphenyl)essigsäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Biphenylacetic acid
A11749.30
A11749.30EA
417
EUR
A11749.30
(4-Biphenyl)essigsäure ≥98%
(4-Biphenyl)essigsäure
Purity: 98%
CAS No.: 5728-52-9
Molecular Formula: C14H12O2
Formula Weight: 212.25
Odour: Odourless
Melting Point: 158-163°
Beilstein Reference: 1211592
Merck Reference: 14,3951
UN No.: 2811
MDL No.: MFCD00004351
Formel:
(C₆H₅C₆H₅)CH₂COOH Molecular Weight: 212,25 g/mol Siedepunkt: 365 °C (1013 hPa) Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004351 CAS-Nummer: 5728-52-9 EINECS: 227-233-2 UN: 2811 ADR: 6.1,III Merck Index: 14,3951 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...