Riboflavin (Vitamin B2) 98

Lieferant: Thermo Fisher Scientific

Synonyme: 6,7-Dimethyl-9-D-ribitylisoalloxazin, Lactoflavin, 7,8-Dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)-benzo[g]pteridin-2,4(3H,10H)-dion, (-)-Riboflavin, Vitamin B2, Vitamin G

A11764.14 A11764.22
A11764.14EA 30.8 EUR
A11764.14 A11764.22
Riboflavin (Vitamin B2) 98
Riboflavin (Vitamin B2)

Purity: 98% CAS No.: 83-88-5 Molecular Formula: C17H20N4O6 Formula Weight: 376.37 Storage Temperature: 2°C to 8°C Physical Form: Powder Odour: Odourless Melting Point: 290° Merck Reference: 14,8200 MDL No.: MFCD00005022


Photoinitiator of polymerization for PAGE gels

Formel: C₁₇H₂₀N₄O₆
Molecular Weight: 376.37 g/mol
Schmelzpunkt: 290 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005022
CAS-Nummer: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR