2-Chlor-3-nitro-5-thiophensulfonylchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A11779.06
A11779.06EA 122 EUR
A11779.06
2-Chlor-3-nitro-5-thiophensulfonylchlorid ≥98%
2-Chlor-3-nitro-5-thiophensulfonylchlorid

Purity: 98% CAS No.: 58457-24-2 Molecular Formula: C4HCl2NO4S2 Formula Weight: 262.09 Melting Point: 52-54° UN No.: 3261 MDL No.: MFCD00067978

Formel: C₄HCl₂NO₄S₂
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067978
CAS-Nummer: 58457-24-2
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR