2,6-Dibromtoluol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11832.03
A11832.06
A11832.03EA
39.1
EUR
A11832.03
A11832.06
2,6-Dibromtoluol ≥98%
2,6-Dibromtoluol
Purity: 98+%
CAS No.: 69321-60-4
Molecular Formula: C7H6Br2
Formula Weight: 249.94
Refractive Index: 1.6060
Melting Point: 2-6°
Boiling Point: 112-113°/7mm
Beilstein Reference: 3235502
MDL No.: MFCD00013524
Formel:
C₇H₆Br₂ Siedepunkt: 112…113 °C (7 mmHg) Schmelzpunkt: 2…6 °C Dichte: 1,812 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013524 CAS-Nummer: 69321-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...