trans-4-Hydroxy-L(-)-prolin ≥99%
Lieferant: Thermo Fisher Scientific
A11851.09
A11851.14
A11851.22
A11851.09EA
46.2
EUR
A11851.09
A11851.14
A11851.22
trans-4-Hydroxy-L(-)-prolin ≥99%
trans-4-Hydroxy-L(-)-prolin
Purity: 99+%
CAS No.: 51-35-4
Molecular Formula: C5H9NO3
Formula Weight: 131.13
Melting Point: 270-273° dec.
Beilstein Reference: 471933
Merck Reference: 14,4840
Optical Rotation: -76° (c=4 in water)
MDL No.: MFCD00064320
Formel:
C₅H₉NO₃ Siedepunkt: 296 °C (1013 hPa) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064320 CAS-Nummer: 51-35-4 EINECS: 200-091-9 Merck Index: 14,4840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...