trans-4-Hydroxy-L(-)-prolin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: L(-)-trans-4-Hydroxyprolin, (S)-(-)-trans-4-Hydroxyprolin, H-Hyp-OH, (2S)-(-)-trans-4-Hydroxypyrrolidin-2-carbonsäure, (S)-(-)-trans-4-Hydroxy-1,3,4,5-tetrahydro-2H-pyrrol- 2-carbonsäure, (S)-(-)-trans-4-Hydroxyazolidin-2-carbonsäure, (S)-(-)-trans-4-Hydroxypyrrolidin-2-carbonsäure

A11851.09 A11851.14 A11851.22
A11851.09EA 46.2 EUR
A11851.09 A11851.14 A11851.22
trans-4-Hydroxy-L(-)-prolin ≥99%
trans-4-Hydroxy-L(-)-prolin

Purity: 99+% CAS No.: 51-35-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 270-273° dec. Beilstein Reference: 471933 Merck Reference: 14,4840 Optical Rotation: -76° (c=4 in water) MDL No.: MFCD00064320

Formel: C₅H₉NO₃
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 270…273 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064320
CAS-Nummer: 51-35-4
EINECS: 200-091-9
Merck Index: 14,4840

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR