Theobromin 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,7-Dimethylxanthin, 3,7-Dimethyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dimethyl-2,6(1H,3H)-purindion, 2,6-Dihydroxy-3,7-dimethylpurin, 3,7-Dimethyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dimethyl-1H,3H-purin-2,6-dion, 3,7-Dihydro-3,7-dimethyl-1H-purin-2,6-dion

A11861.18 A11861.22 A11861.30
A11861.18EA 59.7 EUR
A11861.18 A11861.22 A11861.30
Theobromin 99%
Theobromin

Purity: 99% CAS No.: 83-67-0 Molecular Formula: C7H8N4O2 Formula Weight: 180.17 Melting Point: 345-350° Beilstein Reference: 16464 Merck Reference: 14,9282 MDL No.: MFCD00022830


Phosphosdiesterase inhibitor

Formel: C₇H₈N₄O₂
Molecular Weight: 180.17 g/mol
Schmelzpunkt: 345…350 °C
Dichte: 1.50
Lagertemperatur: Raumtemperatur
MDL: MFCD00022830
CAS-Nummer: 83-67-0
EINECS: 201-494-2
Merck Index: 12,09418

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR