4-Biphenylmethanol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Phenylbenzylalkohol, 4-Hydroxymethylbiphenyl, Biphenyl-4-methanol, (Biphenyl-4-yl)methanol

A11876.09 A11876.18 A11876.30
A11876.09EA 53.6 EUR
A11876.09 A11876.18 A11876.30
4-Biphenylmethanol ≥98%
4-Biphenylmethanol

Purity: 98+% CAS No.: 3597-91-9 Molecular Formula: C13H12O Formula Weight: 184.24 Melting Point: 98-104° Boiling Point: 184°/11mm MDL No.: MFCD00004660

Formel: C₁₃H₁₂O
Siedepunkt: 184 °C (11 mmHg)
Schmelzpunkt: 98…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004660
CAS-Nummer: 3597-91-9
EINECS: 222-745-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR