2-Hydroxy-1,4-naphthochinon 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11880.09
A11880.18
A11880.30
A11880.09EA
30.3
EUR
A11880.09
A11880.18
A11880.30
2-Hydroxy-1,4-naphthochinon 98+%
2-Hydroxy-1,4-naphthochinon
Purity: 98+%
CAS No.: 83-72-7
Molecular Formula: C10H6O3
Formula Weight: 174.16
Melting Point: ca 193° dec.
Beilstein Reference: 1565260
Merck Reference: 14,5393
MDL No.: MFCD00001678
Formel:
C₁₀H₆O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001678 CAS-Nummer: 83-72-7 EINECS: 201-496-3 Merck Index: 14,5393 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...