3-Brom-o-toluidin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline
A11884.14
A11884.14EA
201
EUR
A11884.14
3-Brom-o-toluidin ≥98%
3-Brom-o-toluidin
Purity: 98+%
CAS No.: 55289-36-6
Molecular Formula: C7H8BrN
Formula Weight: 186.06
Flash Point:
Formel:
C₇H₈BrN Molecular Weight: 186,05 g/mol Siedepunkt: 253…255 °C Dichte: 1,510 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051579 CAS-Nummer: 55289-36-6 EINECS: 259-568-5 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...