Aluminiumchlorid, wasserfrei 99%, körnig
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11892.0B
A11892.30
A11892.0E
A11892.0BEA
45.3
EUR
A11892.0B
A11892.30
A11892.0E
Aluminiumchlorid, wasserfrei 99%, körnig
Aluminiumchlorid
Purity: 99%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Odour: Pungent
pH: 2,4
Melting Point: 190° subl.
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Formel:
AlCl₃ Siedepunkt: 180 °C (1013 hPa) Dichte: 2,44 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003422 CAS-Nummer: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II Merck Index: 14,337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...