2'-Methoxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific

A11929.14 A11929.22 AB117448.0025
A11929.14EA 45.7 EUR
A11929.14 A11929.22 ABCRAB117448.0025
2'-Methoxyacetophenon ≥98%
2'-Methoxyacetophenon

Purity: 98% CAS No.: 579-74-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 108°(226°F) Refractive Index: 1.5393 Boiling Point: 119-120°/10mm Beilstein Reference: 1100602 MDL No.: MFCD00008725

Formel: C₉H₁₀O₂
Siedepunkt: 119…120 °C (10 mmHg)
Dichte: 1,090
Flammpunkt: 108 °C (226 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008725
CAS-Nummer: 579-74-8
EINECS: 209-446-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR