2,6-Xylenol ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,6-Dimethylphenol, 2-Hydroxy-3-xylol, 2-Hydroxy-m-xylol, sym.-o-Xylenol
A11947.0E
A11947.36
A11947.0EEA
149
EUR
A11947.0E
A11947.36
2,6-Xylenol ≥99%
2,6-Xylenol
Purity: 99%
CAS No.: 576-26-1
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 78°(172°F)
Melting Point: 44-48°
Boiling Point: 203°
Beilstein Reference: 1446677
Merck Reference: 14,10082
UN No.: 2261
MDL No.: MFCD00002240
Formel:
C₈H₁₀O Molecular Weight: 122.17 g/mol Siedepunkt: 203 °C Schmelzpunkt: 44…48 °C Dichte: 1.15 Flammpunkt: 78 °C (172 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002240 CAS-Nummer: 576-26-1 EINECS: 209-400-1 UN: 2261 ADR: 6.1 II Merck Index: 12,10215 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...