Ethyl-5-bromvalerat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A11952.22 A11952.14 A11952.36
A11952.22EA 173 EUR
A11952.22 A11952.14 A11952.36
Ethyl-5-bromvalerat ≥98%
Ethyl-5-bromvalerat

Purity: 98% CAS No.: 14660-52-7 Molecular Formula: C7H13BrO2 Formula Weight: 209.09 Flash Point: 103°(217°F) Refractive Index: 1.4580 Boiling Point: 227° Beilstein Reference: 1752023 MDL No.: MFCD00000266

Formel: BrH₂C(CH₂)₃COOCH₂CH₃
Siedepunkt: 227 °C
Dichte: 1,306
Flammpunkt: 103 °C (217 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000266
CAS-Nummer: 14660-52-7
EINECS: 238-705-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR