2,5-Dimethyl-2,5-hexandiol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,1,4,4-Tetramethyl-1,4-butanediol
A11984.0E
A11984.22
A11984.36
A11984.0EEA
305
EUR
A11984.0E
A11984.22
A11984.36
2,5-Dimethyl-2,5-hexandiol ≥97%
2,5-Dimethyl-2,5-hexandiol
Purity: 97%
CAS No.: 110-03-2
Molecular Formula: C8H18O2
Formula Weight: 146.23
Flash Point: 126°(258°F)
Odour: Odourless
Boiling Point: 214-215°
Beilstein Reference: 1361437
MDL No.: MFCD00004473
Formel:
C₈H₁₈O₂ Molecular Weight: 146.23 g/mol Siedepunkt: 214…215 °C Schmelzpunkt: 87…91 °C Dichte: 0.898 Flammpunkt: 126 °C (258 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004473 CAS-Nummer: 110-03-2 EINECS: 203-731-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...