3,4-Dimethylbenzamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,4-Dimethylbenzoesäureamid

A11995.03 A11995.06 A11995.14
A11995.03EA 21 EUR
A11995.03 A11995.06 A11995.14
3,4-Dimethylbenzamid ≥98%
3,4-Dimethylbenzamid

Purity: 98% CAS No.: 5580-33-6 Molecular Formula: C9H11NO Formula Weight: 149.19 Melting Point: 101-103° MDL No.: MFCD00052960

Formel: C₉H₁₁NO
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052960
CAS-Nummer: 5580-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR