Tetrapropylammoniumhydrogensulfat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tetrapropylammoniumbisulfat
A12002.14
A12002.14EA
446
EUR
A12002.14
Tetrapropylammoniumhydrogensulfat ≥98%
Tetrapropylammonium hydrogen sulphate
Purity: 98%
CAS No.: 56211-70-2
Molecular Formula: C12H29NO4S
Formula Weight: 283.43
Melting Point: 158-160°
MDL No.: MFCD00038764
Formel:
C₁₂H₂₉NO₄S Molecular Weight: 283.43 g/mol Schmelzpunkt: 158…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038764 CAS-Nummer: 56211-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...